- Dishevelled proteins lead to two signaling pathways. Regulation of LEF-1 and c-Jun N-terminal kinase in mammalian cells.
Dishevelled proteins lead to two signaling pathways. Regulation of LEF-1 and c-Jun N-terminal kinase in mammalian cells.
Dishevelled (Dsh/Dvl) proteins are known to mediate Wnt signaling by up-regulating beta-catenin levels and stimulating T cell factor (TCF)/LEF-1-dependent transcription. We have identified a new Dvl-mediated signaling pathway in that mouse Dvl proteins, when expressed in COS-7 cells, stimulate c-Jun-dependent transcription activity and the kinase activity of the c-Jun N-terminal kinase (JNK). The DEP domain of Dvl1 is essential for JNK activation. By contrast, all three conserved domains of Dvl, including DIX, PDZ, and DEP, are required for up-regulation of beta-catenin and for stimulation of LEF-1-mediated transcription in mammalian cells. Thus, Dvl can lead to two different signaling pathways. Furthermore, the small G proteins of Cdc42 or Rac1, which are involved in JNK activation by many stimuli, do not appear to play a major role in Dvl-mediated JNK activation, because the dominant negative mutants of Cdc42 and Rac1 could not inhibit Dvl-induced JNK activation. This suggests that Dvl may activate JNK via novel pathways.