Merck
  • l-Galactose metabolism in Bacteroides vulgatus from the human gut microbiota.
l-Galactose metabolism in Bacteroides vulgatus from the human gut microbiota.
Biochemistry (2014-06-26)
Merlin Eric Hobbs, Howard J Williams, Brandan Hillerich, Steven C Almo, Frank M Raushel
ABSTRACT
A previously unknown metabolic pathway for the utilization of l-galactose was discovered in a prevalent gut bacterium, Bacteroides vulgatus. The new pathway consists of three previously uncharacterized enzymes that were found to be responsible for the conversion of l-galactose to d-tagaturonate. Bvu0219 (l-galactose dehydrogenase) was determined to oxidize l-galactose to l-galactono-1,5-lactone with kcat and kcat/Km values of 21 s(-1) and 2.0 × 10(5) M(-1) s(-1), respectively. The kinetic product of Bvu0219 is rapidly converted nonenzymatically to the thermodynamically more stable l-galactono-1,4-lactone. Bvu0220 (l-galactono-1,5-lactonase) hydrolyzes both the kinetic and thermodynamic products of Bvu0219 to l-galactonate. However, l-galactono-1,5-lactone is estimated to be hydrolyzed 300-fold faster than its thermodynamically more stable counterpart, l-galactono-1,4-lactone. In the final step of this pathway, Bvu0222 (l-galactonate dehydrogenase) oxidizes l-galactonate to d-tagaturonate with kcat and kcat/Km values of 0.6 s(-1) and 1.7 × 10(4) M(-1) s(-1), respectively. In the reverse direction, d-tagaturonate is reduced to l-galactonate with values of kcat and kcat/Km of 90 s(-1) and 1.6 × 10(5) M(-1) s(-1), respectively. d-Tagaturonate is subsequently converted to d-glyceraldehyde and pyruvate through enzymes encoded within the degradation pathway for d-glucuronate and d-galacturonate.
Sigma-Aldrich
β-Galactose Dehydrogenase from Pseudomonas fluorescens, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥50 units/mg protein (biuret)