Merck
  • An efficient TiO2 coated immobilized system for the degradation studies of herbicide isoproturon: durability studies.
An efficient TiO2 coated immobilized system for the degradation studies of herbicide isoproturon: durability studies.
Chemosphere (2014-05-31)
A Verma, N T Prakash, A P Toor
ABSTRACT
The investigation presents the observations on the use of cement beads for the immobilization of TiO2 for the degradation of herbicide isoproturon. The immobilized system was effective in degrading and mineralizing the herbicide for continuous thirty cycles without losing its durability. Catalyst was characterized by SEM-EDAX for checking the durability of the catalyst. The degradation rate followed first order kinetics as measured by change in absorption intensity in UV range as well as HPLC analysis. Two rounds of TiO2 coating on inert cement beads with average diameter 1.5cm at UV Intensity 25Wm(-2) calcined at 400°C were the optimized conditions for the degradation of herbicide isoproturon. More than 90% TOC and COD reduction along with ammonium ions generation (80%) confirmed the mineralization of isoproturon. Fixed bed baffled reactor studies under solar irradiations using the TiO2 immobilized beads confirmed 85% degradation after 6h. LC-MS studies confirmed the intermediates formation and their subsequent degradation using immobilized system.
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Isoproturon, PESTANAL®, analytical standard
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, wire reel, 200m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire, straight, 1000mm, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 25m, diameter 1.0mm, annealed, 99.6+%
Titanium, wire reel, 20m, diameter 0.125mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 5.53mm, wall thickness 0.41mm, annealed, 99.6+%