Merck
  • Purification and characterization of beta-glucuronidase from Escherichia coli HGU-3, a human intestinal bacterium.
Purification and characterization of beta-glucuronidase from Escherichia coli HGU-3, a human intestinal bacterium.
Biological & pharmaceutical bulletin (1995-09-01)
D H Kim, Y H Jin, E A Jung, M J Han, K Kobashi
ABSTRACT
beta-Glucuronidase was purified 360-fold from Escherichia coli HGU-3, an human intestinal bacterium. The specific activity of the purified enzyme was 17.78 units/mg protein. The enzyme (M.W. 290000) is composed of four subunits (M.W. 72000) with a pI and optimal pH of 4.8 and 6-7, respectively. The apparent Km for p-nitrophenyl-beta-D-glucuronide was found to be 0.22 mM. The enzyme was inhibited by saccharic acid 1,4-lactone, glycyrrhizin, N-ethylmaleimide (NEM) and p-chloromercuriphenylsulfonic acid (PCMS). Using the bile containing bilirubin diglucuronide as a substrate, the purified beta-glucuronidase was able to hydrolyze it to bilirubin. This hydrolyzed bilirubin formed calcium bilirubinate with a reaction mixture containing CaCl2.
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
Pricing and availability is not currently available.