Merck
  • Secondary Organic Aerosol Formation via 2-Methyl-3-buten-2-ol Photooxidation: Evidence of Acid-Catalyzed Reactive Uptake of Epoxides.
Secondary Organic Aerosol Formation via 2-Methyl-3-buten-2-ol Photooxidation: Evidence of Acid-Catalyzed Reactive Uptake of Epoxides.
Environmental science & technology letters (2014-05-27)
Haofei Zhang, Zhenfa Zhang, Tianqu Cui, Ying-Hsuan Lin, Neil A Bhathela, John Ortega, David R Worton, Allen H Goldstein, Alex Guenther, Jose L Jimenez, Avram Gold, Jason D Surratt
ABSTRACT
Secondary organic aerosol (SOA) formation from 2-methyl-3-buten-2-ol (MBO) photooxidation has recently been observed in both field and laboratory studies. Similar to the level of isoprene, the level of MBO-derived SOA increases with elevated aerosol acidity in the absence of nitric oxide; therefore, an epoxide intermediate, (3,3-dimethyloxiran-2-yl)methanol (MBO epoxide), was synthesized and tentatively proposed to explain this enhancement. In this study, the potential of the synthetic MBO epoxide to form SOA via reactive uptake was systematically examined. SOA was observed only in the presence of acidic aerosol. Major SOA constituents, 2,3-dihydroxyisopentanol and MBO-derived organosulfate isomers, were chemically characterized in both laboratory-generated SOA and in ambient fine aerosol collected from the BEACHON-RoMBAS field campaign during the summer of 2011, where MBO emissions are substantial. Our results support the idea that epoxides are potential products of MBO photooxidation leading to the formation of atmospheric SOA and suggest that reactive uptake of epoxides may explain acid enhancement of SOA observed from other biogenic hydrocarbons.
Sigma-Aldrich
2-Methyl-3-buten-1-ol, 98%
Pricing and availability is not currently available.