Merck
  • Studies on the s-cis-trans isomerism for some furan derivatives through IR and NMR spectroscopies and theoretical calculations.
Studies on the s-cis-trans isomerism for some furan derivatives through IR and NMR spectroscopies and theoretical calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-25)
Roberto Rittner, Lucas C Ducati, Cláudio F Tormena, Rodrigo A Cormanich, Barbara C Fiorin, Carolyne B Braga, Raymond J Abraham
ABSTRACT
The s-cis-trans isomerism of two furan derivatives [2-acetyl- (AF) and 2-acetyl-5-methylfuran, (AMF)] was analyzed, using data from the deconvolution of their carbonyl absorption band in two solvents (CH(2)Cl(2) and CH(3)CN). These infrared data showed that the O,O-trans conformers predominate in the less polar solvent (CH(2)Cl(2)), but these equilibria change in a more polar solvent (CH(3)CN) leading to a slight predominance of the O,O-cis conformers, in agreement with the theoretical calculations. The later results were obtained using B3LYP-IEFPCM/6-31++g(3df,3p) level of theory, which taking into account the solvent effects at IEFPCM (Integral Equation Formalism Polarizable Continuum Model). Low temperature (13)CNMR spectra in CD(2)Cl(2) (ca. -75 °C) showed pairs of signals for each carbon, due to the known high energy barrier for the cis-trans interconversion leading to a large predominance of the trans isomers, which decreases in acetone-d(6). This was confirmed by their (1)HNMR spectra at the same temperatures. Moreover, despite the larger hyperconjugative interactions for the O,O-cis isomers, obtained from NBO data, these isomers are destabilized by the their Lewis energy.
Sigma-Aldrich
2-Acetyl-5-methylfuran, 98%