Merck
  • Characterization of the chemical composition of polyisobutylene-based oil-soluble dispersants by fluorescence.
Characterization of the chemical composition of polyisobutylene-based oil-soluble dispersants by fluorescence.
The journal of physical chemistry. B (2014-03-19)
Solmaz Pirouz, Yulin Wang, J Michael Chong, Jean Duhamel
ABSTRACT
A novel methodology based on fluorescence quenching measurements is introduced to determine quantitatively the amine content of polyisobutylene succinimide (PIBSI) dispersants used as engine oil-additives. To this end, a series of five PIBSI dispersants were prepared by reacting 2 mol equiv of polyisobutylene succinic anhydride (PIBSA) with 1 mol equiv of hexamethylenediamine (HMDA), diethylenetriamine, triethylenetetramine, tetraethylenepentamine, and pentaethylenehexamine to yield the corresponding b-PIBSI dispersants. After having demonstrated that the presence of hydrogen bonds between the polyamine linker and the succinimide carbonyls of the dispersants prevents the quantitative analysis of the (1)H NMR and FTIR spectra of the dispersants to determine their chemical composition, alternative procedures based on gel permeation chromatography (GPC) and fluorescence quenching were implemented to estimate the amine content of the b-PIBSI dispersants. Taking advantage of the doubling in size that occurs when 2 mol of PIBSA are reacted with 1 mol of HMDA, a combination of GPC and FTIR was employed to follow how the chemical composition and molecular weight distribution of the polymers produced evolved with the reaction of PIBSA and HMDA mixed at different molar ratios. These experiments provided the PIBSA-to-HMDA molar ratio yielding the largest b-PIBSI dispersants and this molar ratio was then selected to prepare the four other dispersants. Having prepared five b-PIBSI dispersants with well-defined secondary amine content, the fluorescence of the succinimide groups was found to decrease with increasing number of secondary amines present in the polyamine linker. This result suggests that fluorescence quenching provides a valid method to determine the chemical composition of b-PIBSI dispersants which is otherwise difficult to characterize by standard (1)H NMR and FTIR spectroscopies.
Sigma-Aldrich
Pentaethylenehexamine, technical grade
Pricing and availability is not currently available.