Merck
  • Free-operant avoidance behavior by rats after reinforcer revaluation using opioid agonists and D-amphetamine.
Free-operant avoidance behavior by rats after reinforcer revaluation using opioid agonists and D-amphetamine.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-05-03)
Anushka Fernando, Gonzalo Urcelay, Adam Mar, Anthony Dickinson, Trevor Robbins
ABSTRACT
The associative processes that support free-operant instrumental avoidance behavior are still unknown. We used a revaluation procedure to determine whether the performance of an avoidance response is sensitive to the current value of the aversive, negative reinforcer. Rats were trained on an unsignaled, free-operant lever press avoidance paradigm in which each response avoided or escaped shock and produced a 5 s feedback stimulus. The revaluation procedure consisted of noncontingent presentations of the shock in the absence of the lever either paired or unpaired with systemic morphine and in a different cohort with systemic d-amphetamine. Rats were then tested drug free during an extinction test. In both the d-amphetamine and morphine groups, pairing of the drug and shock decreased subsequent avoidance responding during the extinction test, suggesting that avoidance behavior was sensitive to the current incentive value of the aversive negative reinforcer. Experiment 2 used central infusions of D-Ala(2), NMe-Phe(4), Gly-ol(5)]-enkephalin (DAMGO), a mu-opioid receptor agonist, in the periacqueductal gray and nucleus accumbens shell to revalue the shock. Infusions of DAMGO in both regions replicated the effects seen with systemic morphine. These results are the first to demonstrate the impact of revaluation of an aversive reinforcer on avoidance behavior using pharmacological agents, thereby providing potential therapeutic targets for the treatment of avoidance behavior symptomatic of anxiety disorders.
Product Description
Supelco
Morphine sulfate salt solution, 1.0 mg/mL in methanol, drug standard
Sigma-Aldrich
Morphine sulfate salt pentahydrate
Supelco
S(+)-Amphetamine (dextro-Amphetamine) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine for system suitability, European Pharmacopoeia (EP) Reference Standard
Morphine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®