Merck
  • Solid-phase microextraction coupled to high-performance liquid chromatography to determine phenolic compounds in water samples.
Solid-phase microextraction coupled to high-performance liquid chromatography to determine phenolic compounds in water samples.
Journal of chromatography. A (2002-06-13)
A Peñalver, E Pocurull, F Borrull, R M Marcé
ABSTRACT
Solid-phase microextraction (SPME) coupled to high-performance liquid chromatography (HPLC) with ultraviolet (UV) and electrochemical detection (ED) has been applied to determine 11 phenolic compounds considered priority pollutants by the US Environmental Protection Agency. 85 microm polyacrylate fibers were used to extract the analytes from the aqueous samples. Two different designs of the liquid chromatograph were compared in combination with SPME. Dynamic and static modes of desorption in both HPLC designs were compared and the variables affecting both absorption and desorption processes in SPME-HPLC were optimized. Static desorption in both HPLC systems showed better recoveries for the phenolic compounds. The performance of the SPME-HPLC-UV-ED method was evaluated with river water and wastewater samples. The method enabled the determination of phenolic compounds at low levels in these water samples.
Product Description
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga