Merck
  • Palladium-catalyzed allylic alkenylation of allylic alcohols with n-butyl acrylate.
Palladium-catalyzed allylic alkenylation of allylic alcohols with n-butyl acrylate.
Chemical communications (Cambridge, England) (2003-11-01)
Naofumi Tsukada, Tetsuo Sato, Yoshio Inoue
ABSTRACT
Various allylic alcohols reacted with n-butyl acrylate in the presence of p-toluenesulfonic anhydride and palladium catalysts to yield the corresponding n-butyl 2,5-dienoates with high regioselectivity.
Sigma-Aldrich
p-Toluenesulfonic anhydride, 97%