Merck
  • Force-on-Force handgun practice: an intra-individual exploration of stress effects, biomarker regulation, and behavioral changes.
Force-on-Force handgun practice: an intra-individual exploration of stress effects, biomarker regulation, and behavioral changes.
Human factors (2014-04-03)
John Taverniers, Pieter De Boeck
ABSTRACT
The aim of this study was to explore human factors aspects of reality-based "force-on-force" (FoF) handgun practice through a within-subjects field experiment that assessed subjective stress measurements, biomarker regulation, performance outcomes, and behavioral adaptations. FoF handgun practice is a recent training asset for armed officers whereby dynamic opponents may act, react, and even retaliate with specially designed marker ammunition. Predesigned scenarios enable trainees to practice in a simulated real-life environment. A sample of experienced military personnel (N = 20) ran a handgun workshop in two conditions: FoF practice and traditional cardboard-target practice. Intra-individual assessments included anticipated distress, subjective stress, salivary alpha-amylase (sAA), shooting accuracy, and directly observable training seriousness. Compared with the standard cardboard practice condition, FoF exposure caused significant increases in anticipatory distress, subjective stress, and sAA secretion. Furthermore, participants' first encounter with FoF practice (vs. cardboard practice) substantially degraded their shooting performance and had a significant positive impact on the earnestness with which they approached their mission during the workshop. FoF practice is an effective training tool for armed officers because it simulates a realistic work environment by increasing task-specific stress such that it affects important outcomes of professional performance and leads to desirable behavioral changes during training. Potential applications of this research include the introduction of biomarker assessments in human factors research and the design, based on reality-based practice, of effective training procedures for high-reliability professionals.
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid