Merck
  • [Discovery of new structure of class 1 integron in MDR Pseudomonas aeruginosa and its association with drug-resistance].
[Discovery of new structure of class 1 integron in MDR Pseudomonas aeruginosa and its association with drug-resistance].
Wei sheng wu xue bao = Acta microbiologica Sinica (2014-01-01)
Yuying Zhu, Yong Yi, Xi Yang, Na Lv, Jing Li, Baoli Zhu, Yongfei Hu
ABSTRACT
To investigate the characteristics of class I integrons among clinical multidrug-resistant Pseudomonas aeruginosa isolates, and to analyze the association between class I integrons and multidrug resistance. Pseudomonas aeruginosa strains were isolated from clinical samples, and the multidrug-resistant ones were picked out from these isolates. PCR assays were used to detect the variable regions of class I integrons, and the resulting products were then digested with restriction enzyme Sau 3A I and sequenced. SPSS19.0 software was used for statistical analysis. Class I integrons were detected in 27.3% of the clinical multidrug-resistant Pseudomonas aeruginosa isolates. Amplification of variable regions of class I integrons revealed three different gene cassette arrays (1500, 2300 and 4000 bp), two of them were also found in other bacteria. These gene cassette arrays encoded aminoglycoside-modifying enzymes (aadA, aadB, aac(6') II and aadA13), beta-lactamases (blaCARB8 and oxa10) and chloromycetin efflux pump (cmlA8). Analysis results suggested closely relationship between class I integrons and aminoglycoside resistance. We revealed three different gene cassette arrays in the clinical isolates of the multidrug-resistant Pseudomonas aeruginosa, and gene cassette array aadB-aac(6') II -blaCARB8 was the most prevalent. The three gene cassette arrays all contained the aminoglycoside-resistant genes.
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase Blend, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli