Merck
  • A convenient protocol for selective cleavage of 2-hydroxy acid amides. Application to semisynthesis of the cyclic heptapeptide aza HUN-7293.
A convenient protocol for selective cleavage of 2-hydroxy acid amides. Application to semisynthesis of the cyclic heptapeptide aza HUN-7293.
The Journal of organic chemistry (2002-11-26)
Erwin P Schreiner, Michael Kern, Andrea Steck
ABSTRACT
A two-step protocol for the first chemoselective cleavage of 2-hydroxy acid amides has been developed. Mesylation of the model substrate 2-(hydroxypropionylamino)-4-methylpentanoic acid methyl ester (11) followed by treatment with N-ethylthiourea (13) allows cleavage of 2-hydroxy acid amides under smooth conditions. Successful application of this methodology to the open-chain transesterification product 15 (methylester) of the cyclic heptadepsipeptide HUN-7293, a potent inhibitor of inducible cell adhesion molecule expression, delivered the corresponding hexapeptide 18 with unprotected N-terminus in 70-75% yield. This result demonstrates that the protocol developed even works in the presence of an ester and several methylated and unmethylated amide bonds. Finally, a sequence of ligation of methyl D-dehydroglutaminate (20) to the C-terminus of the saponification product 21, followed by the degradation protocol and ring closure, allowed chemical "point mutation" at the DGCN site affording the aza analogue of HUN-7293 (24) in 15% overall yield. To the best of our knowledge this is the first report on chemoselective cleavage of 2-hydroxy acid amides.
Sigma-Aldrich
N-Ethylthiourea, 99%