Merck
  • Highly dispersed noble-metal/chromia (core/shell) nanoparticles as efficient hydrogen evolution promoters for photocatalytic overall water splitting under visible light.
Highly dispersed noble-metal/chromia (core/shell) nanoparticles as efficient hydrogen evolution promoters for photocatalytic overall water splitting under visible light.
Nanoscale (2010-07-21)
Naoyuki Sakamoto, Hajime Ohtsuka, Takahiro Ikeda, Kazuhiko Maeda, Daling Lu, Masayuki Kanehara, Kentaro Teramura, Toshiharu Teranishi, Kazunari Domen
ABSTRACT
Highly dispersed rhodium nanoparticles (1.7 +/- 0.3 nm) prepared by a liquid-phase reduction method were loaded on a solid solution of GaN and ZnO without forming aggregates, achieving improved activity for visible-light-driven overall water splitting when the nanoparticles are coated with a chromia shell.
Sigma-Aldrich
Sodium 3-mercapto-1-propanesulfonate, technical grade, 90%