Merck
  • Split-dose administration of a dual-action, low-volume bowel cleanser for colonoscopy: the SEE CLEAR I study.
Split-dose administration of a dual-action, low-volume bowel cleanser for colonoscopy: the SEE CLEAR I study.
Gastrointestinal endoscopy (2013-04-10)
Douglas K Rex, Philip O Katz, Gerald Bertiger, Stephen Vanner, Lawrence C Hookey, Vivian Alderfer, Raymond E Joseph
ABSTRACT
New bowel cleansers for colonoscopy that lead to improved efficacy, safety, and tolerability are needed. This study evaluated a nonphosphate, dual-action, low-volume, orange-flavored preparation containing sodium picosulfate and magnesium citrate (P/MC). Multicenter, assessor-blinded, randomized, noninferiority study. University hospitals, academic medical centers, and private clinics across the United States. Adults preparing for colonoscopy. P/MC versus 2 L of polyethylene glycol solution (2L PEG-3350) and two 5-mg bisacodyl tablets. This phase 3 study investigated the efficacy, safety, and tolerability of split-dose administration of P/MC versus day-before dosing of 2L PEG-3350 and two 5-mg bisacodyl tablets (SEE CLEAR I study). Efficacy was evaluated by using the Aronchick and Ottawa scales; noninferiority and superiority analyses were performed. Safety was assessed by monitoring adverse events (AEs). Tolerability was measured via a patient questionnaire. The intent-to-treat population consisted of 601 patients who self-administered P/MC (n = 304) or 2L PEG-3350 and bisacodyl tablets (n = 297). P/MC was superior to 2L PEG-3350 and bisacodyl tablets in overall colon cleansing (84.2% vs 74.4%; 1-sided 97.5% confidence interval [CI], 3.4) (Aronchick scores of excellent or good) and in cleansing of the ascending (89.5% vs 78.8%; 1-sided 97.5% CI, 4.9), mid (transverse and descending) (92.4% vs 85.9%; 1-sided 97.5% CI, 1.6), and rectosigmoid (92.4% vs 87.2%; 1-sided 97.5% CI, 0.4) segments of the colon (Ottawa scores of excellent, good, or fair). Commonly reported AEs related to the bowel preparations were nausea, vomiting, headache, and chills. Patient-reported tolerability, including ease of consumption and taste, was significantly higher for P/MC than 2L PEG-3350 and bisacodyl tablets (P < .0001). Because of differences in administration and volume of the bowel preparations, the study was designed to be a single-assessor, blinded study. The bowel-cleansing effects and patient acceptability of split-dose P/MC were superior to day-before dosing with 2L PEG-3350 and bisacodyl tablets.
Product Description
Picosulfate for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.