Merck
  • A case of drowning whilst under the influence of brotizolam, flunitrazepam and ethanol.
A case of drowning whilst under the influence of brotizolam, flunitrazepam and ethanol.
Soudni lekarstvi (2011-03-19)
Naoko Tanaka, Hiroshi Kinoshita, Mostofa Jamal, Eriko Ohkubo, Mitsuru Kumihashi, Kiyoshi Ameno
ABSTRACT
A case of drowning involving brotizolam, flunitrazepam and ethanol ingestion was presented. Quantitative toxicological analysis showed that the concentrations of brotizolam, 7-aminoflunitrazepam (a metabolite of flunitrazepam) and ethanol in the femoral blood were 0.025 microg/ml, 0.094 microg/ml and 0.29 mg/ml, respectively, and these drugs were also detected in the stomach contents. We concluded that the cause of death was drowning whilst under the influence of combined use of brotizolam, flunitrazepam and ethanol.
Product Description
Brotizolam, European Pharmacopoeia (EP) Reference Standard