Merck
  • Preventive strategy for Candida gut translocation during ischemia-reperfusion injury supervening on protein-calorie malnutrition.
Preventive strategy for Candida gut translocation during ischemia-reperfusion injury supervening on protein-calorie malnutrition.
Chinese journal of digestive diseases (2006-01-18)
F Marotta, R Barreto, S Kawakita, E Minelli, K Pavasuthipaisit, A Lorenzetti, M Nishiwaki, F Gelosa, E Fesce, R Okura
ABSTRACT
The aim of the present study was to test the hypothesis that protein-calorie malnutrition aggravates the gut translocation of Candida albicans triggered by mesenteric ischemia-reperfusion (IR) injury in an experimental model while testing a natural product containing the antifungal anethole/polygodial mixture (Kolorex). MFI strain white mice (n = 90) were randomly allocated to a 4-week dietary regimen: (1) standard pellet diet containing 25% casein; (2) low-protein (2.5%) casein diet; (3) as group 2 plus oral supplementation with 20 microL of a 5% solution of Kolorex during the last 4 days. Twenty rats from each of these groups (termed 1a, 2a and 3a) were orally inoculated with Candida suspension 6 h prior to mesenteric IR injury. Animals of each group but without Candida inoculation (termed 1b, 2b and 3b) served as control. A colon permeability study was carried out as well. Rats were killed prior to the IR injury and 3 h afterwards. Control rats were killed at the same time. Over 60% of the mesenteric lymph nodes and 30% of kidney samples were positive for C. albicans in the low-protein-fed rats after IR injury. Kolorex significantly decreased that rate of positivity and also significantly reduced the concentration of C. albicans per gram of each positive tissue sample examined. Protein-calorie malnourished animals showed a statistically significant increase in colon permeability and this phenomenon further increased after IR injury. The groups of rats treated with Kolorex compound showed a partial, although significant, improvement of this parameter. These results suggest that Kolorex might exert a competitive effect against with C. albicans colonization. The present study represents the first experimental in vivo investigation of the anethole/polygodial-containing compound under the specific conditions of calorie-protein malnutrition and the results have potential clinical interest.
Sigma-Aldrich
Polygodial, ≥97% (HPLC)