Merck
  • Involvement of calmodulin and protein kinase C in cholecystokinin release by bombesin from STC-1 cells.
Involvement of calmodulin and protein kinase C in cholecystokinin release by bombesin from STC-1 cells.
Pancreas (2000-10-20)
A Takahashi, S Tanaka, Y Miwa, H Yoshida, A Ikegami, J Niikawa, K Mitamura
ABSTRACT
The mouse intestinal neuroendocrine tumor cell line STC-1 secretes cholecystokinin (CCK) and other hormones. We investigated the role of Ca2+, calmodulin (CaM), and protein kinase C (PKC) in the regulation of CCK release from STC-1 cells. Phorbol 12-myristate 13-acetate (TPA) significantly stimulated CCK release. Staurosporine significantly inhibited CCK release from STC-1 cells stimulated by TPA in a dose-dependent manner. The absence of extracellular calcium completely inhibited CCK release from TPA-stimulated STC-1 cells. Neurotensin did not stimulate CCK release from these cells. W-7, a CaM antagonist, reduced CCK release from STC-1 cells stimulated by bombesin in a dose-dependent manner. These findings suggest that CaM and PKC play an important role in the regulation of CCK release from STC-1 cells stimulated by bombesin.
Sigma-Aldrich
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride, ≥98% (HPLC)