Merck
  • Postmenstrual age and CYP2D6 polymorphisms determine tramadol o-demethylation in critically ill neonates and infants.
Postmenstrual age and CYP2D6 polymorphisms determine tramadol o-demethylation in critically ill neonates and infants.
Pediatric research (2008-03-05)
Karel Allegaert, Ron H N van Schaik, Steve Vermeersch, Rene Verbesselt, Veerle Cossey, Christine Vanhole, Marianne van Fessem, Jan de Hoon, John N van den Anker
ABSTRACT
To document determinants of O-demethylation in critically ill (pre)term neonates and infants, tramadol (M) and O-demethyl tramadol (M1) concentrations were quantified in eighty-six 24 h urine collections and 168 plasma samples. A significant correlation of urine log M/M1 (0.98, SD 0.66) and plasma log M/M1 (0.78, SD 0.45) with postmenstrual age (PMA) (r = -0.69 and -0.65) was observed. One-way analysis of variance documented a significant decrease in urine log and plasma log M/M1 with increasing CYP2D6 activity score (F value 11.6 and 22.55). PMA and CYP2D6 activity score determined the urine and plasma log M/M1 (R 0.59 and 0.64) in a forward multiple regression model. We therefore conclude that PMA and CYP2D6 polymorphisms determined O-demethylation activity in (pre)term neonates and young infants, illustrating the impact of pharmacogenetics on drug metabolism in neonates although a relevant part of the interindividual varaibility remained unexplained. Besides compound-specific relevance, CYP2D6 iso-enzyme specific data on in vivo ontogeny of O-demethylation can contribute to safer and more effective administration of drugs metabolized by the same route in this population.