Merck
  • Involvement of MAPK signaling pathway in the osteogenic gene expressions of Cervi Pantotrichum Cornu in MG-63 human osteoblast-like cells.
Involvement of MAPK signaling pathway in the osteogenic gene expressions of Cervi Pantotrichum Cornu in MG-63 human osteoblast-like cells.
Life sciences (2013-11-19)
Jee Hyun Kim, Tae Young Jung, Jungchul Seo, Sena Lee, Myung-Gyou Kim, Kang-Hyun Leem, Sung Chul Lim
ABSTRACT
The purposes of this study were to determine whether Cervi Pantotrichum Cornu (CPC) has osteogenic activities in human osteoblastic MG-63 cells and to investigate the underlying molecular mechanism. The effects of CPC on alkaline phosphatase activity, collagen synthesis, and calcium deposits were measured. The COL1A1, ALPL, BGLAP, and SPP1 expressions were measured by real-time PCR. Phosphorylated MAP kinases (ERK1/2, JNK1/2, p38, ELK1, and cJUN) were studied by western blot analysis. The involvement of MAPK pathway in osteogenic gene expressions was determined by using each selective MAPK inhibitor (PD98059, SP600125, and SB203580). CPC increased alkaline phosphatase activity, collagen synthesis, and calcium deposits. CPC activated ERK1/2, JNK1/2, p38, and ELK1 phosphorylation except cJUN. CPC increased the COL1A1, ALPL, BGLAP, and SPP1 gene expressions. The elevated COL1A1 and BGLAP expressions were inhibited by PD98059, SP600125 or SB203580. The elevated ALPL expression was blocked by SB203580. The elevated SPP1 expression was inhibited by SP600125 or SB203580. CPC increased COL1A1 and BGLAP expressions via ERK1/2, JNK1/2, and p38 MAPKs pathways and SPP1 expression via JNK1/2 and p38 pathways. p38 pathway is needed for ALPL expression. These results imply that MAPK signaling pathway is an indispensable factor for bone matrix genes expression of CPC in MG-63 human osteoblast-like cells.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein