Merck
  • Effect of α-linolenic acid on streptozotocin-induced diabetic retinopathy indices in vivo.
Effect of α-linolenic acid on streptozotocin-induced diabetic retinopathy indices in vivo.
Archives of medical research (2013-10-15)
Jun-hui Shen, Qi Ma, Sheng-rong Shen, Shen-grong Shen, Guo-Tong Xu, Undurti N Das
ABSTRACT
Both oxidative stress and inflammation play a significant role in the pathobiology of diabetic retinopathy. Increased consumption of polyunsaturated fatty acids (PUFAs) may prevent or postpone the occurrence of diabetic retinopathy. Hence, the effect of α-linolenic acid (ALA), an essential fatty acid, on oxidative stress, inflammatory indices and production of vascular endothelial growth factor (VEGF) in streptozotocin-induced diabetic retinopathy indices in vivo was studied. Serum and retina concentrations of vascular endothelial growth factor (VEGF), brain-derived neurotrophic factor (BDNF), interleukin-6 (IL-6), plasma and retina concentrations of lipid peroxides and antioxidant enzymes were estimated in streptozotocin (STZ)-induced diabetic animals. STZ-induced diabetic rats had significantly higher levels of VEGF in the serum and retina and IL-6 in the serum, whereas BDNF was lower in the serum, all of which reverted to near normal in ALA-treated diabetic animals. STZ treatment decreased serum glutathione peroxidase levels, which was restored to normal by both pre- and post-ALA treatment groups. STZ-induced changes in serum glutathione peroxidase, BDNF, VEGF and IL-6 that reverted to near control by ALA treatment, especially in ALA + STZ group, lending support to the concept that both oxidative stress and inflammation participate in DR and ALA treatment is of benefit in its prevention.
Product Description
Supelco
Linolenic acid, analytical standard
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Sigma-Aldrich
Linolenic acid, ≥99%