Merck
  • Astrocyte control of synaptic NMDA receptors contributes to the progressive development of temporal lobe epilepsy.
Astrocyte control of synaptic NMDA receptors contributes to the progressive development of temporal lobe epilepsy.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-09)
Jerome Clasadonte, Jinghui Dong, Dustin J Hines, Philip G Haydon
ABSTRACT
Astrocytes modulate neuronal activity, synaptic transmission, and behavior by releasing chemical transmitters in a process termed gliotransmission. Whether this process impacts epilepsy in vivo is not known. We show that genetic impairment of transmitter release from astrocytes by the expression of a glial dominant-negative SNARE domain in mice reduced epileptiform activity in situ, delayed seizure onset after pilocarpine-induced status epilepticus, and attenuated subsequent progressive increase in seizure frequency in vivo. The reduced seizure frequency was accompanied by attenuation of hippocampal damage and behavioral deficits. As the delay in seizure onset and the reduced seizure frequency were mimicked by intracerebroventricular delivery of the NMDA receptor (NMDAR) antagonist D-(-)-2-amino-5-phosphonopentanoate in WT littermates and because dominant-negative SNARE expression leads to a hypofunction of synaptic NMDARs, we conclude that astrocytes modulate epileptogenesis, recurrent spontaneous seizures, and pathophysiological consequences of epilepsy through a pathway involving NMDARs.
Product Description
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Pilocarpine nitrate salt, ≥98% (HPLC)
Pilocarpine nitrate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine nitrate, meets USP testing specifications