Merck
  • Comparison of levofloxacin versus moxifloxacin for multidrug-resistant tuberculosis.
Comparison of levofloxacin versus moxifloxacin for multidrug-resistant tuberculosis.
American journal of respiratory and critical care medicine (2013-08-10)
Won-Jung Koh, Seung Heon Lee, Young Ae Kang, Chang-Hoon Lee, Jae Chol Choi, Jae Ho Lee, Seung Hun Jang, Kwang Ha Yoo, Ki Hwan Jung, Ki Uk Kim, Sang Bong Choi, Yon Ju Ryu, Kyung Chan Kim, Soojung Um, Yong Soo Kwon, Yee Hyung Kim, Won-Il Choi, Kyeongman Jeon, Yong Il Hwang, Se Joong Kim, Young Seok Lee, Eun Young Heo, Juhee Lee, Yeo Woon Ki, Tae Sun Shim, Jae-Joon Yim
ABSTRACT
Levofloxacin (LFX) and moxifloxacin (MXF) are the two most frequently recommended fluoroquinolones for treatment of patients with multidrug-resistant tuberculosis (MDR-TB). However, studies comparing the effectiveness of LFX and MXF among patients with MDR-TB are lacking. To compare the effectiveness of LFX and MXF in terms of culture conversion after 3 months of treatment for MDR-TB. In this prospective multicenter randomized open label trial, we randomly assigned 182 patients with MDR-TB (sensitive to LFX and MXF) to receive either LFX (750 mg/day; 90 patients) or MXF (400 mg/day; 92 patients) with a background drug regimen. The primary outcome was the proportion of patients who achieved sputum culture conversion at 3 months of treatment. Secondary outcomes were time to culture conversion and time to smear conversion, with data censored at 3 months, and the proportions of adverse drug reactions. At 3 months of treatment, 68 (88.3%) of the 77 patients in the LFX group and 67 (90.5%) of the 74 in the MXF group showed conversion to negative sputum cultures (odds ratio for LFX compared with MXF, 0.78; 95% confidence interval, 0.27-2.20). Adverse drug reactions were reported in six patients (7.7%) in the LFX group and four (5.2%) in the MXF group (P = 0.75). The choice of LFX or MXF for treatment of patients with MDR-TB may not affect sputum culture conversion at 3 months of treatment. Clinical trial registered with www.clinicaltrials.gov (NCT 01055145).
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Levofloxacin, analytical standard