- The first total synthesis of (+/-)-linderol A, a tricyclic hexahydrodibenzofuran constituent of Lindera umbellata bark, with potent inhibitory activity on melanin biosynthesis of cultured B-16 melanoma cells.
The first total synthesis of (+/-)-linderol A, a tricyclic hexahydrodibenzofuran constituent of Lindera umbellata bark, with potent inhibitory activity on melanin biosynthesis of cultured B-16 melanoma cells.
Organic letters (2001-05-12)
M Yamashita, N Ohta, I Kawasaki, S Ohta
PMID11348234
ABSTRACT
[reaction in text] The first total synthesis of (+/-)-linderol A, a hexahydrodibenzofuran isolated from Lindera umbellata bark, with potent inhibitory activity on melanin biosynthesis of cultured B-16 melanoma cells was achieved via a 20-step of reaction in 7.64% overall yield starting from 4,6-dimethoxysalicylaldehyde.