Merck
  • THE 1-DEOXY-D-XYLULOSE-5-PHOSPHATE PATHWAY OF ISOPRENOID BIOSYNTHESIS IN PLANTS.
THE 1-DEOXY-D-XYLULOSE-5-PHOSPHATE PATHWAY OF ISOPRENOID BIOSYNTHESIS IN PLANTS.
Annual review of plant physiology and plant molecular biology (2004-03-12)
Hartmut K. Lichtenthaler
ABSTRACT
In plants the biosynthesis of prenyllipids and isoprenoids proceeds via two independent pathways: (a) the cytosolic classical acetate/mevalonate pathway for the biosynthesis of sterols, sesquiterpenes, triterpenoids; and (b) the alternative, non-mevalonate 1-deoxy-d-xylulose-5-phosphate (DOXP) pathway for the biosynthesis of plastidic isoprenoids, such as carotenoids, phytol (a side-chain of chlorophylls), plastoquinone-9, isoprene, mono-, and diterpenes. Both pathways form the active C5-unit isopentenyl diphosphate (IPP) as the precursor from which all other isoprenoids are formed via head-to-tail addition. This review summarizes current knowledge of the novel 1-deoxy-d-xylulose-5-phosphate (DOXP) pathway for isopentenyl diphosphate biosynthesis, apparently located in plastids. The DOXP pathway of IPP formation starts from D-glyceraldehyde-3-phosphate (GA-3-P) and pyruvate, with DOXP-synthase as the starting enzyme. This pathway provides new insight into the regulation of chloroplast metabolism.
Sigma-Aldrich
1-Deoxy-D-xylulose-5-phosphate sodium salt, ≥99.0% (TLC)