Merck
  • Patterns of cerebrospinal fluid catechols support increased central noradrenergic responsiveness in aging and Alzheimer's disease.
Patterns of cerebrospinal fluid catechols support increased central noradrenergic responsiveness in aging and Alzheimer's disease.
Biological psychiatry (1999-09-24)
M A Raskind, E R Peskind, C Holmes, D S Goldstein
ABSTRACT
High cerebrospinal fluid (CSF) norepinephrine (NE) concentrations in aging and Alzheimer's disease (AD) could reflect decreased NE clearance from central nervous system (CNS) extracellular fluid or increased release of NE into CNS extracellular fluid. Measuring CSF concentrations of the intraneuronal NE metabolite dihydroxyphenylglycol (DHPG), an estimate of NE clearance, and the NE precursor dihydroxyphenylacetic acid (DOPA), an estimate of NE biosynthesis, can help differentiate these mechanisms. NE, DHPG, and DOPA were determined by HPLC in CSF and plasma obtained following yohimbine, clonidine, and placebo. Ten AD, 10 older, and 11 young subjects were studied. CSF DOPA following yohimbine was higher in older and AD than in young subjects. CSF DHPG did not differ among groups. Plasma DOPA following yohimbine was higher in AD than in young subjects. During alpha-2 adrenoreceptor blockade in both aging and AD, there are increased responses of CNS NE biosynthesis and release with unchanged CNS NE clearance. This pattern is consistent with partial loss of CNS noradrenergic neurons with compensatory activation of remaining CNS noradrenergic neurons. Given the marked loss of locus coeruleus (LC) noradrenergic neurons in AD, achievement of high CSF NE suggests particularly prominent compensatory activation of remaining LC neurons in this disorder.
Product Description
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis