Merck
  • The purification of human urinary kallikrein with ion-exchange radial flow membrane chromatography.
The purification of human urinary kallikrein with ion-exchange radial flow membrane chromatography.
Biomedical chromatography : BMC (1996-05-01)
H Wang, T Li, H Zou, Y Zhang, J Chao, L Chao
ABSTRACT
A method was developed for the purification of kallikrein from human urine. The procedure consisted of three steps: ultradialysis, diethyl-(2-hydroxypropyl) aminoethyl (QAE) ion exchange radial flow membrane chromatography and affinity chromatography on aprotinin agarose. It is simple and suitable for large-scale purification. The purified product was checked by SDS-PAGE and matrix-assisted laser desorption:ionization mass spectrometery (MALDI). A single band with apparent molecular weight (MW) 42,000 in SDS-PAGE and a single monomer peak with MW33,000 in MALDI were observed, respectively. The biological activity tested by ELISA showed positive immunological identity of the purified product compared with human urinary kallikrein standard.
Sigma-Aldrich
Pro-Phe-Arg-7-amido-4-methylcoumarin, ≥95%