Merck
Synthesis of 15 alpha-hydroxyestrogen 15-N-acetylglucosaminides.
Steroids (1995-03-01)
E Suzuki, S Namba, H Kurihara, J Goto, Y Matsuki, T Nambara
ABSTRACT
The synthesis of 15-N-acetylglucosaminides of 15 alpha-hydroxyesterone, 15 alpha-hydroxyestradiol, and 15 alpha-hydroxyestriol (estetrol) is described. The latter two were prepared by condensation of 2-acetamido-1 alpha-chloro-1,2-dideoxy-3,4,6-trio-O-acetyl-D-glucopyranose with appropriately protected 15 alpha-hydroxyestrogens by the Koenigs-Knorr reaction employing cadmium carbonate as a catalyst. Subsequent removal of protecting groups with methanolic potassium hydroxide provided the desired conjugates. 15 alpha-Hydroxyestrone 15-N-acetylglucosaminide was synthesized from the corresponding 15 alpha-hydroxyestradiol derivative by Jones oxidation followed by brief alkaline hydrolysis. These conjugates underwent enzymatic hydrolysis with beta-N-acetylglucosaminidase from Jack beans to produce 15 alpha-hydroxyestrogens.
Sigma-Aldrich
Cadmium carbonate, powder, 98%