Merck
  • N-Mannich-base prodrugs of 5-iodo-2'-deoxycytidine as topical delivery enhancers.
N-Mannich-base prodrugs of 5-iodo-2'-deoxycytidine as topical delivery enhancers.
Pharmaceutical research (1987-08-01)
S A Koch, K B Sloan
ABSTRACT
Two Mannich-base prodrugs of 5-iodo-2'-deoxycytidine (5-IDC) have been synthesized. The prodrugs exhibit increased lipid solubility compared to 5-IDC and rapidly revert to 5-IDC in buffer. One of the prodrugs delivered about twice as much 5-IDC from isopropyl myristate (IPM) through hairless mouse skin in diffusion-cell experiments as did 5-IDC from IPM. Subsequent applications of theophylline/propylene glycol onto the diffusion cells to determine the effect of prodrug/IPM, 5-IDC/IPM, or IPM on the resistance of the skins to subsequent applications showed that the prodrug/IPM had no more effect than IPM itself.