Merck
  • Reaction of 2-amino-2-deoxy-D-glucose and lysine: isolation and characterization of 2,5-bis(tetrahydroxybutyl)pyrazine.
Reaction of 2-amino-2-deoxy-D-glucose and lysine: isolation and characterization of 2,5-bis(tetrahydroxybutyl)pyrazine.
Carbohydrate research (1988-12-31)
G Candiano, G M Ghiggeri, R Gusmano, L Zetta, E Benfenati, G Icardi
ABSTRACT
The reaction of 2-amino-2-deoxy-D-glucose with lysine in water under simulated physiological conditions gave several browning products, with characteristic optical (lambda max 340 nm) and fluorescent properties (emission at 430 nm for excitation at 362 nm). The major product was isolated and characterized by mass spectrometry and n.m.r. spectroscopy as 2,5-bis(tetrahydroxybutyl)pyrazine derived by the condensation of two molecules of 2-amino-2-deoxy-D-glucose.