Merck
Analog applications of photochemical switches.
Advanced materials (Deerfield Beach, Fla.) (2013-02-22)
Graeme Copley, Thomas A Moore, Ana L Moore, Devens Gust
ABSTRACT
Molecules that change their structure in response to a stimulus such as light or an added chemical can act as molecular switches. Such switches can be chemically linked to other active moieties to create molecular "devices" for various purposes. There has been much activity of late in the use of molecular switches such as photochromes in the construction of molecular logic gates that carry out binary or digital functions. However, ensembles of such molecules can also act as analog devices. Here, examples of a molecular photonic signal transducer and two mimics of photosynthetic photoregulatory processes are discussed.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Anthracene, analytical standard