Merck
  • High-pressure-promoted Diels-Alder approach to biaryls: application to the synthesis of the cannabinols family.
High-pressure-promoted Diels-Alder approach to biaryls: application to the synthesis of the cannabinols family.
The Journal of organic chemistry (2012-08-24)
Lucio Minuti, Andrea Temperini, Eleonora Ballerini
ABSTRACT
Diels-Alder reactions of a range of 1-(alkoxy/alkyl/halogen-substituted phenyl)buta-1,3-dienes with methyl propiolate carried out in a green ethanolic medium under 9 kbar pressure were investigated. The use of high pressure as activating method of the Diels-Alder reactions allows efficient and regioselective generation of a series of cyclohexadienyl-benzene cycloadducts that are oxidized to the corresponding biaryls. The alkoxy/alkyl/halogen-substituted biaryls produced are useful precursors for accessing substituted 6H-benzo[c]chromen-6-ones and the cannabinols family.
Product Description
Supelco
Cannabinol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®