Merck
  • Pimavanserin, a 5-HT2A receptor inverse agonist, reverses psychosis-like behaviors in a rodent model of Alzheimer's disease.
Pimavanserin, a 5-HT2A receptor inverse agonist, reverses psychosis-like behaviors in a rodent model of Alzheimer's disease.
Behavioural pharmacology (2012-07-04)
Diana L Price, Douglas W Bonhaus, Krista McFarland
ABSTRACT
Alzheimer's disease (AD) is a neurodegenerative disorder characterized by a progressive deterioration in cognitive functioning. Overall, 25-50% of patients with AD also show symptoms of psychosis including hallucinations and delusions. As all available antipsychotic drugs have a 'black-box' warning for use in these patients because of increased mortality, no appropriate treatment for psychotic symptoms in AD currently exists. In the present study, we examined whether selective antagonism of 5-HT(2A) serotonin receptors has antipsychotic-like activity in an animal model of AD. Mice receiving an intracerebroventricular infusion of the amyloid β(25-35) peptide fragment showed AD-like histopathology and a psychosis-related behavioral phenotype with enhanced responses to the psychostimulants 2,5-dimethoxy-4-iodoamphetamine hydrochloride and amphetamine as well as disrupted prepulse inhibition. Treatment with pimavanserin, a selective serotonin 5-HT(2A) receptor inverse agonist, prevented 2,5-dimethoxy-4-iodoamphetamine hydrochloride-induced head twitches, reversed the augmented locomotor response to amphetamine, and normalized prepulse inhibition in mice with amyloid pathology. These data suggest that an infusion of amyloid β might induce alterations in serotonergic function that underlie a psychosis-like phenotype that can be normalized by treatment with a 5-HT(2A) inverse agonist. This in turn suggests that 5-HT(2A) inverse agonists, such as pimavanserin, might have therapeutic benefits in the treatment of psychosis in AD patients.