- Selected antimicrobial essential oils eradicate Pseudomonas spp. and Staphylococcus aureus biofilms.
Selected antimicrobial essential oils eradicate Pseudomonas spp. and Staphylococcus aureus biofilms.
Applied and environmental microbiology (2012-04-03)
Nicole L Kavanaugh, Katharina Ribbeck
PMID22467497
ABSTRACT
Biofilms are difficult to eliminate with standard antimicrobial treatments due to their high antibiotic resistance relative to free-living cells. Here, we show that selected antimicrobial essential oils can eradicate bacteria within biofilms with higher efficiency than certain important antibiotics, making them interesting candidates for the treatment of biofilms.
MATERIALS