Merck
  • N,N'-dicyclohexylsulfamide and N,N'-diphenethylsulfamide are anticonvulsant sulfamides with affinity for the benzodiazepine binding site of the GABA(A) receptor and anxiolytic activity in mice.
N,N'-dicyclohexylsulfamide and N,N'-diphenethylsulfamide are anticonvulsant sulfamides with affinity for the benzodiazepine binding site of the GABA(A) receptor and anxiolytic activity in mice.
Biochemical pharmacology (2011-11-08)
Cristina Wasowski, Luciana Gavernet, Ivana A Barrios, Maria L Villalba, Valentina Pastore, Gisela Samaja, Andrea Enrique, Luis E Bruno-Blanch, Mariel Marder
ABSTRACT
A set of sulfamides designed, synthesized and evaluated against maximal electroshock seizure (MES) and pentilenetetrazol (PTZ) tests with promising results, were tested for their affinity for the benzodiazepine binding site of the GABA(A) receptor. The most active compounds, N,N'-dicyclohexylsulfamide (7) and N,N'-diphenethylsulfamide (10), competitively inhibited the binding of [(3)H]-flunitrazepam to the benzodiazepine binding site with K(i)±SEM values of 27.7±4.5μM (n=3) and 6.0±1.2μM (n=3), respectively. The behavioral actions of these sulfamides, i.p. administered in mice, were examined in the plus-maze, hole-board and locomotor activity assays. Compound 7 exhibited anxiolytic-like effects in mice evidenced by a significant increase of the parameters measured in the hole-board test (at 1 and 3mg/kg) and the plus-maze assay (at 1 and 3mg/kg). Compound 10 evidenced anxiolytic activity in the plus-maze and the hole-board tests at 1mg/kg. Locomotor activity of mice was not modified by compound 7 or 10 at the doses tested. Flumazenil, a non selective benzodiazepine binding site antagonist, was able to completely reverse the anxiolytic-like effects of these sulfamides, proving that the GABA(A) receptor is implicated in this action. Anxiety represents a major problem for people with epilepsy. The use of anxiolytic and anticonvulsant sulfamides would be beneficial to individuals who suffer from both disorders.
Product Description
Flunitrazepam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Flunitrazepam
Supelco
Flunitrazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®