Merck
  • The site of intestinal disappearance of DL-methionine and methionine hydroxy analog differs in pigs.
The site of intestinal disappearance of DL-methionine and methionine hydroxy analog differs in pigs.
Journal of animal science (2011-01-11)
J A Jendza, P A Geraert, D Ragland, O Adeola
ABSTRACT
A study was conducted with pigs to determine the sites of and the influence of dietary wheat middlings (WM) as a fiber source on the absorption of dl-Met (DLM) and dl-2-hydroxy-4-methylthio butanoic acid (HMTBA). Twelve 25-kg barrows were used in a replicated 6 × 4 incomplete Latin-square design with 6 diets and 4 periods per pig. Each pig was surgically fitted with simple T-cannulas in the terminal duodenum and ileum 2 wk before the initiation of the experiment and housed in a stainless-steel metabolism crate. The 6 diets were arranged in a 2 × 3 factorial of WM at 0 (CON) or 8% and Met source including none, DLM, or HMTBA. Supplemental DLM doubled the duodenal free Met concentration from 9.6 to 19.6 g/100 g of DMI. Supplemental DLM also increased the apparent ileal digestibility (AID) of free Met from 52 to 92% (P < 0.05). Adding 8% WM increased (P < 0.01) duodenal NDF concentrations by 2 g/100 g of DMI and decreased (P < 0.05) AID of free Met. No HMTBA was detected in digesta collected from the duodenum or ileum of pigs fed the CON + HMTBA or the WM + HMTBA diets, indicating complete absorption before the terminal duodenum regardless of WM inclusion. Interactions between Met supplementation and WM were observed for the digestibility of all other nutrients (P < 0.05). Fiber and AA digestibility in the ileum were the greatest for the CON + HMTBA diet, and least for the WM + HMTBA diet. The CON + HMTBA diet had greater AID of ADF, NDF, CP, and Thr, and greater AID of ADF, NDF, ash, Arg, Ile, Gly, and Pro when compared with the CON and CON + DLM diets, respectively (P < 0.05). The WM + HMTBA had less AID of ADF, CP, and all AA with the exception of Lys, Trp, and Val when compared with the CON + HMTBA (P < 0.05). In summary, absorption of HMTBA in the pig is complete by the end of the duodenum, but negative interaction between HMTBA and WM can decrease the AID of most AA.
Sigma-Aldrich
2-Hydroxy-4-(methylthio)butyric acid calcium salt, ≥99.0% (NT)