Merck
  • Central and peripheral anti-inflammatory effects of maprotiline on carrageenan-induced paw edema in rats.
Central and peripheral anti-inflammatory effects of maprotiline on carrageenan-induced paw edema in rats.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2010-06-25)
V Hajhashemi, H Sadeghi, M Minaiyan, A Movahedian, A Talebi
ABSTRACT
To explore the site of action of maprotiline, as an atypical antidepressant, on carrageenan-induced paw edema. Male Wistar rats were used. Firstly, the anti-inflammatory effect of systemic maprotiline (12.5, 25 and 50 mg kg(-1)) was assessed using a paw edema model. Secondly, different doses of maprotiline were administrated intracerebroventricularly, intrathecally and locally before carrageenan challenge. Finally, we tried to reverse the anti-inflammatory effect of maprotiline by propranolol (10 mg kg(-1)), prazosin (4 mg kg(-1)), yohimbine (10 mg kg(-1)), naloxone (4 mg kg(-1)) and mifepristone (5 mg kg(-1)). Systemic, intracerebroventricular and subplantar application of maprotiline significantly inhibited peripheral edema, but intrathecal maprotiline did not alter the degree of paw swelling. The applied antagonists failed to change the anti-inflammatory activity of maprotiline. These results demonstrate that maprotiline has a potent anti-inflammatory effect and this effect is linked to the peripheral and supraspinal actions of the drug.
Sigma-Aldrich
Maprotiline hydrochloride, >99% (HPLC), powder
Supelco
Maprotiline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®