Merck
  • Clinical value of circulating endothelial cells and circulating tumor cells in metastatic breast cancer patients treated first line with bevacizumab and chemotherapy.
Clinical value of circulating endothelial cells and circulating tumor cells in metastatic breast cancer patients treated first line with bevacizumab and chemotherapy.
Annals of oncology : official journal of the European Society for Medical Oncology (2010-03-18)
F-C Bidard, C Mathiot, A Degeorges, M-C Etienne-Grimaldi, R Delva, X Pivot, C Veyret, L Bergougnoux, P de Cremoux, G Milano, J-Y Pierga
ABSTRACT
We investigated whether circulating tumor cells (CTCs) and circulating endothelial cells (CECs) predict clinical outcome of first-line chemotherapy combined with bevacizumab in metastatic breast cancer patients. In a French substudy of the MO19391 trial, CTC and CEC counts (CellSearch system) at baseline and changes after two cycles of treatment were correlated with time to progression (TtP). CTC and CEC levels were not correlated in the 67 patients included. At baseline, CTC positivity was a significant prognostic marker for TtP at a threshold of 3 CTC/7.5 ml (P < 0.05) but not at 5 CTC/7.5 ml (P = 0.09). Baseline CEC levels (median 17 CEC/4 ml, range 1-769) were associated with age > or =45 years (P = 0.01), elevated lactate dehydrogenase (P < 0.01) and not with TtP at any threshold. Changes of CTC count during treatment were not a surrogate of TtP, with any of the model tested (threshold based or relative decrease in percent). However, increase in CEC count was associated with improved TtP, at the threshold of 20 CEC/4 ml (P < 0.01). Bevacizumab combined with first-line chemotherapy may modify the predictive value of CTC during treatment possibly due to impaired tumor cells intravasation through vessels endothelium. Variations in CEC levels appear to be a promising early surrogate marker of TtP under antiangiogenic treatment.
Product Description
Docetaxel trihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)