Merck
  • Substance P promotes susceptibility to Pseudomonas aeruginosa keratitis in resistant mice: anti-inflammatory mediators downregulated.
Substance P promotes susceptibility to Pseudomonas aeruginosa keratitis in resistant mice: anti-inflammatory mediators downregulated.
Investigative ophthalmology & visual science (2008-04-04)
Sharon A McClellan, Yunfan Zhang, Ronald P Barrett, Linda D Hazlett
ABSTRACT
Studies have shown that blocking substance P (SP) binding to neurokinin 1 receptor with spantide I prevents Pseudomonas aeruginosa-induced corneal perforation in susceptible C57BL/6 mice. This study tested the effect of SP injection on the resistance response (cornea heals) of BALB/c mice. The day before infection, mice were injected intraperitoneally with SP or PBS. Disease was graded by clinical score, slit lamp, plate count, real-time RT-PCR, and ELISA assays, and polymorphonuclear neutrophils (PMNs) were quantitated using a myeloperoxidase assay. In additional experiments, BALB/c mice were injected intraperitoneally with vasoactive intestinal peptide (VIP) antagonist and similarly analyzed. Mice injected with SP exhibited worsened disease on days 1 to 7 after infection compared with controls. SP injection resulted in elevated PMN levels and viable bacterial counts in the cornea 3 and 5 days after infection. mRNA expression for NFkappaB and type 1 cytokines (e.g., IFN-gamma), as well as for TNF-alpha, MIP-2, IL-18, IL-6, and IL-1beta, were significantly elevated, whereas VIP and cytokines TGF-beta and IL-10 were significantly reduced. Differences in mRNA expression were selectively confirmed at the protein level by ELISA for NFkappaB, IL-1beta, and IL-10. VIP antagonist treatment also resulted in exacerbated disease scores, elevated proinflammatory mediators, and reduced anti-inflammatory mediators. These data provide evidence that the neuropeptide SP, among its broad systemic effects, is a potent neuroimmunoregulator that promotes susceptibility in the resistant BALB/c mouse by overcoming the anti-inflammatory effects of VIP and IL-10 and that a balance between SP and VIP levels may be critical in disease resolution.