Merck
  • Atropisomeric property of 1-(2,6-difluorobenzyl)-3-[(2R)-amino-2-phenethyl]-5-(2-fluoro-3-methoxyphenyl)-6-methyluracil.
Atropisomeric property of 1-(2,6-difluorobenzyl)-3-[(2R)-amino-2-phenethyl]-5-(2-fluoro-3-methoxyphenyl)-6-methyluracil.
Chirality (2005-10-01)
Fabio C Tucci, Tao Hu, Michael F Mesleh, Allan Bokser, Eric Allsopp, Timothy D Gross, Zhiqiang Guo, Yun-Fei Zhu, R Scott Struthers, Nicholas Ling, Chen Chen
ABSTRACT
1-(2,6-Difluorobenzyl)-3-[(2R)-amino-2-phenethyl]-5-(2-fluoro-3-methoxyphenyl)-6-methyluracil (6), a potent and orally active antagonist of the human gonadotropin-releasing hormone receptor, exists as a pair of atropisomers in solution, which was detected by NMR spectroscopy, and separable by HPLC. In addition to a (R)-configured benzylamine, there is a second stereogenic element due to the presence of a chiral axis between the substituted 5-phenyl group and the uracil core. The rate constant of the interconversion (k = 5.07 x 10(-5) s(-1)) of these two atropisomers was determined by proton NMR analysis of a diastereoisomer-enriched sample in aqueous solution at 25 degrees C, and the corresponding Gibbs free energy DeltaG(#) of rotation barrier (97.4 kJ mol(-1)) was calculated using the Eyring equation. The diastereoisomer half-life at physiological temperature (37 degrees C) in aqueous media was estimated to be about 46 min.
Sigma-Aldrich
6-Methyluracil, 97%