Merck
  • Processing of metallurgical residues by flotation - bench-scale studies on two industrial products.
Processing of metallurgical residues by flotation - bench-scale studies on two industrial products.
Waste management (New York, N.Y.) (2005-06-25)
S R Rao, J A Finch
ABSTRACT
Resource recovery from two metallurgical residues by flotation was investigated applying an electrostatic model to select initial conditions. The first, a sulphation roast/water leach residue, was processed to float lead sulphate, comparing dodecylamine and xanthate collectors. From the second, a neutralization residue, gypsum, was recovered by reverse flotation of ferric hydroxide, comparing oleate and sulphonate collectors. In both cases, further upgrading by acid leaching was considered.
Sigma-Aldrich
Lead(II) sulfate, 98%
Sigma-Aldrich
Lead(II) sulfate, 99.995% trace metals basis