Merck
  • [TIC4]endomorphins, analogues of endomorphins, have significantly enhanced vasorelaxant effects in rat aorta rings.
[TIC4]endomorphins, analogues of endomorphins, have significantly enhanced vasorelaxant effects in rat aorta rings.
Protein and peptide letters (2005-05-24)
Qian-Yu Zhao, Qiang Chen, Yun Feng, Xin Lin, Rui Wang
ABSTRACT
[Tic(4)]EM1 and [Tic(4)]EM2, new endomorphins (EMs) analogues, caused relaxation of rat aorta rings precontracted with phenylphrine in a concentration-dependent manner and were 240- to 370-fold more potent than EMs. This effect was inhibited by endothelium removal or by incubation with NO synthase inhibitor L-NNA or opioid receptor antagonist naloxone. The results demonstrate that [Tic(4)]EMs have NO- and endothelium-dependent vasorelaxant effects which are mediated by the opioid receptor.
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%