Merck
  • Medium-size droplets of methyl ricinoleate are reduced by cell-surface activity in the gamma-decalactone production by Yarrowia lipolytica.
Medium-size droplets of methyl ricinoleate are reduced by cell-surface activity in the gamma-decalactone production by Yarrowia lipolytica.
Letters in applied microbiology (2000-04-04)
Y Waché, K Bergmark, J L Courthaudon, M Aguedo, J M Nicaud, J M Belin
ABSTRACT
Size of methyl ricinoleate droplets during biotransformation into gamma-decalactone by Yarrowia lipolytica was measured in both homogenized and non-homogenized media. In non-homogenized but shaken medium, droplets had an average volume surface diameter d32 of 2.5 microm whereas it was 0.7 microm in homogenized and shaken medium. But as soon as yeast cells were inoculated, both diameters became similar at about 0.7 microm and did not vary significantly until the end of the culture. The growth of Y. lipolytica in both media was very similar except for the lag phase which was lowered in homogenized medium conditions.
Sigma-Aldrich
Methyl ricinoleate, ≥99% (GC)
Supelco
Methyl ricinoleate, analytical standard