Merck
Toxicity of field bindweed (Convolvulus arvensis) to mice.
Veterinary and human toxicology (1995-10-01)
P C Schultheiss, A P Knight, J L Traub-Dargatz, F G Todd, F R Stermitz
ABSTRACT
The effects of feeding high and low doses of field bindweed (Convolvulus arvensis) to mice were investigated. Bindweed contains several alkaloids, including pseudotropine, and lesser amounts of tropine, tropinone, and meso-cuscohygrine. Mice fed bindweed exclusively died or were euthanized after 4-7 d and had severe hepatic necrosis and gastritis with ulceration or erosions. Mice fed low doses of bindweed along with standard laboratory mouse diet for 6 or 8 w had no clinical disease or gross lesions on necropsy examination but did have histologic lesions of mild multifocal hepatitis and gastritis.
Sigma-Aldrich
Tropine, ≥97.0% (NT)
Sigma-Aldrich
Tropinone, 99%