Merck
  • Effects of amphetamine and amfonelic acid on the disposition of striatal newly synthesized dopamine.
Effects of amphetamine and amfonelic acid on the disposition of striatal newly synthesized dopamine.
European journal of pharmacology (1982-02-19)
H H Miller, P A Shore
ABSTRACT
A comparison of the in vivo biochemical actions of the psychotomimetic central stimulants, d-amphetamine (d-AMPH) and amfonelic acid (AFA), on the metabolism of rat striatal newly synthesized [3H]dopamine (DA) was made by pulse labeling with [3H]tyrosine. No evidence for the formation of the alcoholic DA metabolites [3H]3-methoxy-4-hydroxyphenylethanol (MOPET) or [3H]3,4-dihydroxyphenylethanol (DOPET) was found in control or drug-treated animals. Both [3H]3,4-dihydroxyphenylacetic acid (DOPAC) and [3H]homovanillic acid (HVA) concentrations were increased by AFA in the presence of haloperidol, while [3H]DA content was decreased. In contrast, d-AMPH, in the presence of haloperidol, decreased [3H]DOPAC and increased [3H]DA, even in monoamine oxidase-blocked rats. Thus monoamine oxidase inhibition did not appear to be a major factor in the action of amphetamine to increase [3H]DA, but cannot be excluded as a contributing factor to the lowering of [3H]DOPAC. Similar actions of d-AMPH were seen on preformed DA. Amphetamine may release newly synthesized DA in such a way that some of the released DA enters the neuronal storage system.
Sigma-Aldrich
Homovanillyl alcohol, 99%
Pricing and availability is not currently available.