Merck
  • Properties, in theory and practice, of novel gel filtration media for standard liquid chromatography.
Properties, in theory and practice, of novel gel filtration media for standard liquid chromatography.
Journal of chromatography (1989-08-04)
L Hagel, H Lundström, T Andersson, H Lindblom
ABSTRACT
The influence of experimental parameters on the separation result in gel filtration may readily be predicted from a few basic equations, as demonstrated with the aid of experimental observations on Sephacryl HR. Excellent agreement between the predicted resolution and that determined experimentally was found for parameters such as temperature, particle size, column length, flow velocity, sample volume and gel porosity. The theoretical prediction of an optimum sample volume for a constant processing rate was also experimentally verified. An exhaustive investigation of the physical, chemical and functional properties of Sephacryl HR was undertaken to facilitate the interpretation of experimental observations.
Sigma-Aldrich
Sephacryl®, 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 400-HR, MW range 20-8000 kDa (globular proteins)
Sigma-Aldrich
Sephacryl®, 100-HR, MW range 1000-100,000 Da (globular proteins)
Sigma-Aldrich
Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 500-HR, MW range 40-20,000 kDa (dextrans)