Merck
  • [Pyridazinilides (XIV). The action of p-phenyl-phenacyl bromide on 3-phenyl-pyridazine].
[Pyridazinilides (XIV). The action of p-phenyl-phenacyl bromide on 3-phenyl-pyridazine].
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (1989-01-01)
M Caproşu, M Ungureanu, M Pavelescu, C Radu, M Petrovanu
ABSTRACT
This time the synthesis of some new derivatives obtained by the quaternary substitution of one of the heterocyclic nitrogen atoms and by reactions of dipolar cycloaddition is presented. The results of the tests on their antimicrobial and antiinflammatory actions are discussed.