Merck
  • Supramolecular hydrogel based on graphene oxides for controlled release system.
Supramolecular hydrogel based on graphene oxides for controlled release system.
Journal of nanoscience and nanotechnology (2013-05-08)
Qian-Yi Cheng, Bao-Hang Han
ABSTRACT
Supramolecular hydrogel drug delivery system that composed of F127 modified small-sized graphene oxide (F127-SGO) and alpha-CD was prepared and the controlled release behavior was studied using doxorubicin hydrochloride (DOX) as a model drug. As compared with native hydrogel formed from F127 and alpha-CD, the SGO-containing hybrid hydrogel system shows multiple binding sites to load drug molecules and a more controllable release process that facilitates to tune the drug delivery system. These properties make the supramolecular hydrogel a potential candidate for controlled drug delivery system.
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, flakes