Merck
Worsening of contact dermatitis by oral hydroxyzine: a case report.
Dermatology online journal (2013-03-12)
Nianda Reid, Sunit Jariwala, Golda Hudes, Jason Fodeman, Elina Jerschow, David Rosenstreich
ABSTRACT
Hydroxyzine is commonly used to treat pruritic skin lesions. Although rare, hydroxyzine can sometimes be linked to worsening dermatitis in patients who have sensitivities to phenothiazines and/or ethylenediamines. Herein we describe a patient who developed papulovesicular eruptions following the use of topical neomycin. Our patient's contact dermatitis initially improved after the use of oral steroids. However, the patient's skin condition was exacerbated by the continued use of hydroxyzine to treat her pruritus. Patch testing was positive at 48 hours for neomycin sulfate, ethylenediamine dihydrochloride, and p-phenylenediamine. Given the suspected cross-reactivity between hydroxyzine and ethylenediamine, hydroxyzine was discontinued and the patient's cutaneous symptoms improved. In summary, physicians must be aware that oral hydroxyzine can worsen contact dermatitis in ethylenediamine-sensitive patients.
Product Description
Supelco
Neomycin trisulfate salt hydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Neomycin sulphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydroxyzine dihydrochloride, ≥97%