Merck
  • Effect of TiO2 photocatalytic activity in a HDPE-based food packaging on the structural and microbiological stability of a short-ripened cheese.
Effect of TiO2 photocatalytic activity in a HDPE-based food packaging on the structural and microbiological stability of a short-ripened cheese.
Food chemistry (2013-02-16)
Matteo Gumiero, Donatella Peressini, Andrea Pizzariello, Alessandro Sensidoni, Lucilla Iacumin, Giuseppe Comi, Rosanna Toniolo
ABSTRACT
A high density polyethylene (HDPE)/calcium carbonate (CaCO(3)) film containing TiO(2) was prepared via blown film extrusion process. The photocatalytic properties of this film were evaluated by voltammetric, UV-Vis spectrophotometric and gas chromatographic measurements following the decomposition rate of suitably selected molecular probes, such as 4-hydroxybenzoic acid and methylene blue. The film containing 1% w/w of TiO(2) displayed a profitable and reproducible photoinduced degradation activity towards target organic compounds. The effect of packaging photocatalytic activity on the structural and microbiological stability of a short-ripened cheese was studied. Cheese structure was assessed by dynamic, small deformation rheological tests. A container consisting of a multilayer material, where the layer brought in contact with the food, made from the HDPE+CaCO(3)+TiO(2) composite matrix, was able to provide a greater maintenance of the original cheese structure than a rigid container currently used, mainly due to the inhibition of lactic acid bacteria and coliforms.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Polyethylene (LDPE), ERM®, certified reference material